Assay Details
Target Gene Details
Entrez Gene ID: | 66136 |
Gene Name: | zinc ribbon domain containing, 1 |
Gene Aliases: |
1110014N07Rik, Rpa12 |
Location: |
Chr.17:36954349-36958520 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Znrd1 | NM_023162.4 | NP_075651.1 | ||
AF230339.1 | AAG50161.1 | |||
AK003714.1 | BAB22954.1 | |||
AK005976.1 | BAB24350.1 | |||
AK019152.1 | ||||
BC043016.1 | AAH43016.1 |
More Information
Set Membership: |
Intragenic Non-exonic |