Assay Details
Target Gene Details
Entrez Gene ID: | 224826 |
Gene Name: | ubiquitin protein ligase E3 component n-recognin 2 |
Gene Aliases: |
9930021A08Rik, AI462103, AW540746, E130209G04Rik, mKIAA0349 |
Location: |
Chr.17:46928291-47010775 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 40 - Exon 40 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ubr2 | NM_001177374.1 | NP_001170845.1 | ||
NM_146078.3 | NP_666190.2 | |||
AK078002.1 | ||||
AK122254.2 | BAC65536.3 | |||
AK143202.1 | BAE25301.1 | |||
AK147445.1 | BAE27917.1 | |||
AY061885.1 | AAL32102.1 | |||
AY280958.1 | AAQ17202.1 | |||
BC025617.1 | AAH25617.1 | |||
BC026391.1 | AAH26391.1 | |||
BC031403.1 | AAH31403.1 | |||
BC075642.1 | AAH75642.1 |
More Information
Set Membership: |
Intragenic Non-exonic |