Assay Details
Target Gene Details
Entrez Gene ID: | 108077 |
Gene Name: | superkiller viralicidic activity 2-like (S. cerevisiae) |
Gene Aliases: |
4930534J06Rik, AW214248, Ddx13, SKI, Ski2w |
Location: |
Chr.17:34839226-34850204 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Skiv2l | NM_021337.2 | 20 | 2449 | NP_067312.2 |
AK029763.1 | 20 | 2438 | BAC26604.1 | |
AK087810.1 | 16 | 3083 | BAC40010.1 | |
AK139051.1 | 19 | 2523 | ||
AK159854.1 | 20 | 2450 | BAE35430.1 | |
AK169864.1 | 20 | 2420 | BAE41420.1 | |
AK178949.1 | ||||
AK180219.1 | ||||
AK185511.1 | ||||
AK202117.1 | ||||
AK204941.1 | ||||
AK205255.1 | ||||
AK206945.1 | ||||
AK214376.1 | ||||
AK216744.1 | ||||
BC023478.1 | 8 | 1053 | AAH23478.1 | |
BC065999.1 | 20 | 2421 | AAH65999.1 |
More Information
Set Membership: |
Intragenic Exonic |