Assay Details
Target Gene Details
Entrez Gene ID: | 108013 |
Gene Name: | CUGBP, Elav-like family member 4 |
Gene Aliases: |
A230070D14Rik, BRUNOL-4, Brul4, Brunol4, C130060B05Rik |
Location: |
Chr.18:25477620-25754162 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Celf4 | NM_001146292.1 | NP_001139764.1 | ||
NM_001146293.1 | NP_001139765.1 | |||
NM_001146294.1 | NP_001139766.1 | |||
NM_001174074.1 | NP_001167545.1 | |||
NM_133195.3 | NP_573458.2 | |||
AF314173.1 | AAK00298.1 | |||
AF515450.1 | AAO22167.1 | |||
AK048427.1 | BAC33334.1 | |||
AK051458.1 | BAC34649.1 | |||
BC048405.1 | AAH48405.1 | |||
BC052744.1 | AAH52744.1 |