Assay Details
Target Gene Details
Entrez Gene ID: | 71302 |
Gene Name: | Rho GTPase activating protein 26 |
Gene Aliases: |
1810044B20Rik, 2610010G17Rik, 4933432P15Rik, AI853435, mKIAA0621 |
Location: |
Chr.18:38992968-39376285 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arhgap26 | NM_175164.4 | NP_780373.3 | ||
AK129176.1 | BAC97986.1 | |||
BC141394.1 | ||||
BM935209.1 |
More Information
Set Membership: |
Intragenic Non-exonic |