Assay Details
Target Gene Details
Entrez Gene ID: | 72238 |
Gene Name: | TBC1 domain family, member 5 |
Gene Aliases: |
1600014N05Rik |
Location: |
Chr.17:50733124-51180198 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tbc1d5 | NM_001285991.1 | NP_001272920.1 | ||
NM_001285993.1 | NP_001272922.1 | |||
NM_028162.4 | NP_082438.3 | |||
AK005461.1 | BAB24052.1 | |||
AK033710.1 | BAC28439.1 | |||
AK144467.1 | BAE25904.1 | |||
AK154975.1 | BAE32965.1 | |||
AK163366.1 | ||||
AK220251.1 | BAD90176.1 | |||
BC043113.1 | AAH43113.1 | |||
BC098328.1 | AAH98328.1 |
More Information
Set Membership: |
Intragenic Non-exonic |