Assay Details
Target Gene Details
Entrez Gene ID: | 381073 |
Gene Name: | neuropeptide W |
Gene Aliases: |
Gm935 |
Location: |
Chr.17:24657330-24658425 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Npw | NM_001099664.1 | NP_001093134.1 | ||
AK131881.1 | BAE20848.1 |