Assay Details
Target Gene Details
Entrez Gene ID: | 71461 |
Gene Name: | PTK7 protein tyrosine kinase 7 |
Gene Aliases: |
8430404F20Rik, chz, mPTK7/CCK4 |
Location: |
Chr.17:46564451-46630049 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptk7 | NM_175168.4 | 19 | 3128 | NP_780377.1 |
AB055408.1 | 5 | 627 | BAC53806.1 | |
AF531873.1 | 19 | 2978 | AAQ10071.1 | |
AK018379.1 | BAE43251.1 | |||
AK053044.1 | 19 | 3061 | BAC35248.1 | |
AK163135.1 | 19 | 3061 | BAE37208.1 | |
BC027800.1 | 8 | 1183 | AAH27800.1 | |
BC076578.1 | 19 | 3128 | AAH76578.1 |
More Information
Set Membership: |
Intragenic Exonic |