Assay Details
Target Gene Details
Entrez Gene ID: | 14979 |
Gene Name: | H2-K region expressed gene 6 |
Gene Aliases: |
D17H6S112E, H-2Ke6, Hsd17b8, Ke-6, Ke6, Ring2 |
Location: |
Chr.17:34026033-34028055 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
H2-Ke6 | NM_013543.2 | 2 | 165 | NP_038571.2 |
BC086927.1 | 2 | 165 | AAH86927.1 | |
CX243612.1 | 2 | 134 | ||
KJ650217.1 | 2 | 157 |
More Information
Set Membership: |
Intragenic Exonic |