Assay Details
Target Gene Details
Entrez Gene ID: | 20016 |
Gene Name: | polymerase (RNA) I polypeptide C |
Gene Aliases: |
40kDa, AA409007, AA959927, AL024089, Polr1e, RPA40, Rpo1-1 |
Location: |
Chr.17:46243920-46248045 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Polr1c | NM_009085.2 | 3 | 277 | NP_033111.2 |
AK132114.1 | 3 | 219 | BAE20986.1 | |
AK146240.1 | 3 | 227 | BAE27005.1 | |
BC130018.1 | 3 | 278 | ||
BY267923.1 | 3 | 277 |
More Information
Set Membership: |
Intragenic Exonic |