Assay Details
Target Gene Details
Entrez Gene ID: | 18044 |
Gene Name: | nuclear transcription factor-Y alpha |
Gene Aliases: |
AA407810, Cbf-b, SEZ-10, SEZ10 |
Location: |
Chr.17:48386885-48409901 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nfya | NM_001110832.1 | NP_001104302.1 | ||
NM_010913.2 | NP_035043.1 | |||
AK004729.1 | BAB23511.1 | |||
AK143723.1 | BAE25515.1 | |||
AK156487.1 | BAE33729.1 | |||
BC029695.1 | AAH29695.1 | |||
BC057099.1 | AAH57099.1 | |||
M86214.1 | AAA39816.1 | |||
M86215.1 | AAA39817.1 | |||
X55315.1 | CAA39023.1 |
More Information
Set Membership: |
Intragenic Non-exonic |