Assay Details
Target Gene Details
Entrez Gene ID: | 98053 |
Gene Name: | general transcription factor IIF, polypeptide 1 |
Gene Aliases: |
2810405L04Rik, C76800, RAP74 |
Location: |
Chr.17:57003401-57011393 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gtf2f1 | NM_133801.2 | 1 | 88 | NP_598562.1 |
AK013002.1 | 1 | 102 | BAB28588.1 | |
AK078048.1 | 1 | 101 | BAC37108.1 | |
AK168236.1 | 1 | 101 | BAE40186.1 | |
AK168243.1 | 1 | 101 | BAE40193.1 | |
BC023081.1 | 1 | 88 | AAH23081.1 | |
BC031123.1 | 1 | 218 | AAH31123.1 |
More Information
Set Membership: |
Intragenic Exonic |