Assay Details
Target Gene Details
Entrez Gene ID: | 78249 |
Gene Name: | adhesion G protein-coupled receptor F4 |
Gene Aliases: |
4632435A09Rik, AV239010, Gpr115 |
Location: |
Chr.17:42656887-42692284 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adgrf4 | NM_001289499.1 | 4 | 383 | NP_001276428.1 |
NM_001289500.1 | 4 | 370 | NP_001276429.1 | |
NM_001289501.1 | 4 | 414 | NP_001276430.1 | |
NM_030067.2 | 4 | 457 | NP_084343.1 | |
AK019508.1 | 4 | 460 | BAB31767.1 | |
BC089564.1 | 4 | 383 | AAH89564.1 | |
BC140980.1 | 4 | 323 | ||
BY714064.1 | 4 | 458 |