Assay Details
Target Gene Details
Entrez Gene ID: | 435529 |
Gene Name: | adhesion G protein-coupled receptor F2 |
Gene Aliases: |
Gpr111, PGR20 |
Location: |
Chr.17:42694028-42749145 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adgrf2 | NM_001033493.2 | NP_001028665.2 | ||
AK142836.1 | BAE25206.1 | |||
BB662982.1 | ||||
BC141353.1 |