Assay Details
Target Gene Details
Entrez Gene ID: | 69310 |
Gene Name: | PARK2 co-regulated |
Gene Aliases: |
1700008H23Rik |
Location: |
Chr.17:10403012-10840191 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pacrg | NM_027032.2 | NP_081308.1 | ||
AK005771.1 | BAB24230.1 | |||
AK135510.1 | BAE22559.1 | |||
BC120740.1 |