Assay Details
Target Gene Details
Entrez Gene ID: | 353344 |
Gene Name: | opsin 5 |
Gene Aliases: |
Gpr136, Neuropsin, PGR12, TMEM13 |
Location: |
Chr.17:42556779-42620470 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Opn5 | NM_181753.4 | NP_861418.2 | ||
AY288426.1 | AAP72135.1 | |||
AY318865.1 | AAR08201.1 | |||
BC125611.1 | ||||
BC132010.1 |
More Information
Set Membership: |
Intragenic Non-exonic |