Assay Details
Target Gene Details
Entrez Gene ID: | 16004 |
Gene Name: | insulin-like growth factor 2 receptor |
Gene Aliases: |
AI661837, CD222, CI-MPR, M6P/IGF2R, Mpr300 |
Location: |
Chr.17:12682406-12769706 on Build GRCm38 |
Assay Gene Location: | Within Exon 31 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Igf2r | NM_010515.2 | 31 | 4502 | NP_034645.2 |
AK088699.1 | 6 | 728 | BAC40512.1 | |
BC150817.1 | 31 | 4413 | ||
BC171973.1 | 31 | 4413 | ||
CD545656.1 | 2 | 141 | ||
CN461279.1 | 5 | 551 | ||
U04710.1 | 31 | 4458 | AAA19568.1 |
More Information
Set Membership: |
Intragenic Exonic |