Assay Details
Target Gene Details
Entrez Gene ID: | 224938 |
Gene Name: | praja 2, RING-H2 motif containing |
Gene Aliases: |
AI447901, AL022700, Neurodap1, mKIAA0438 |
Location: |
Chr.17:64281006-64331883 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pja2 | NM_001025309.1 | 9 | 2116 | NP_001020480.1 |
NM_144859.2 | 8 | 1930 | NP_659108.1 | |
AF493070.1 | 9 | 2173 | AAO85470.1 | |
AK122282.1 | 9 | 2075 | BAC65564.1 | |
AK144586.1 | 9 | 2103 | BAE25949.1 | |
AK168371.1 | 8 | 1912 | BAE40303.1 | |
BC004742.1 | 6 | 960 | AAH04742.1 | |
BC017130.1 | 7 | 1856 | AAH17130.1 |
More Information
Set Membership: |
Intragenic Exonic |