Assay Details
Target Gene Details
Entrez Gene ID: | 224938 |
Gene Name: | praja 2, RING-H2 motif containing |
Gene Aliases: |
AI447901, AL022700, Neurodap1, mKIAA0438 |
Location: |
Chr.17:64281006-64331883 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pja2 | NM_001025309.1 | NP_001020480.1 | ||
NM_144859.2 | NP_659108.1 | |||
AF493070.1 | AAO85470.1 | |||
AK122282.1 | BAC65564.1 | |||
AK144586.1 | BAE25949.1 | |||
AK164025.1 | ||||
AK168371.1 | BAE40303.1 | |||
BY065517.1 |
More Information
Set Membership: |
Intragenic Non-exonic |