Assay Details
Target Gene Details
Entrez Gene ID: | 20112 |
Gene Name: | ribosomal protein S6 kinase, polypeptide 2 |
Gene Aliases: |
90kDa, D17Wsu134e, Rps6ka-rs1, Rsk3, p90rsk, pp90rsk |
Location: |
Chr.17:7088975-7303315 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rps6ka2 | NM_011299.4 | NP_035429.1 | ||
AJ131021.1 | CAB44492.1 | |||
AK019881.1 | BAB31901.1 | |||
AK132079.1 | BAE20973.1 | |||
BC043064.1 | AAH43064.1 | |||
BC051079.1 | AAH51079.1 | |||
BC055331.1 | AAH55331.1 | |||
BC056946.1 | AAH56946.1 |
More Information
Set Membership: |
Intragenic Non-exonic |