Assay Details
Target Gene Details
Entrez Gene ID: | 12458 |
Gene Name: | chemokine (C-C motif) receptor 6 |
Gene Aliases: |
CC-CKR-6, CCR-6, Cmkbr6, KY411 |
Location: |
Chr.17:8236043-8257141 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccr6 | NM_001190333.1 | NP_001177262.1 | ||
NM_009835.4 | NP_033965.1 | |||
AB016031.1 | BAA82443.1 | |||
AK089712.1 | BAC40946.1 |