Assay Details
Target Gene Details
Entrez Gene ID: | 224997 |
Gene Name: | discs, large (Drosophila) homolog-associated protein 1 |
Gene Aliases: |
4933422O14Rik, 9630002F18, AI845682, AI848168, BB075781, D17Bwg0511e, GKAP/SAPAP, GKPA/SAPAP, Gkap, Sapap1, mKIAA4162 |
Location: |
Chr.17:69969078-70821413 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dlgap1 | NM_001128180.1 | 3 | 461 | NP_001121652.1 |
NM_001128181.1 | 3 | 433 | NP_001121653.1 | |
NM_027712.3 | 4 | 1448 | NP_081988.3 | |
NM_177639.6 | 4 | 1448 | NP_808307.2 | |
AK031410.1 | 8 | 2138 | BAC27391.1 | |
AK035763.1 | 7 | 1999 | ||
AK136926.1 | 3 | 394 | BAE23171.1 | |
AK147448.1 | 8 | 2265 | BAE27919.1 | |
AK161941.1 | 3 | 436 | BAE36644.1 | |
AK163430.1 | 3 | 630 | ||
AK163689.1 | 3 | 442 | BAE37458.1 | |
AK220515.1 | 7 | 2328 | BAD90519.1 | |
AY243846.2 | 3 | 1283 | AAP70755.2 | |
BC062120.1 | 4 | 1449 | AAH62120.1 | |
BC094369.1 | 3 | 657 | AAH94369.1 |
More Information
Set Membership: |
Intragenic Exonic |