Assay Details
Target Gene Details
Entrez Gene ID: | 224997 |
Gene Name: | discs, large (Drosophila) homolog-associated protein 1 |
Gene Aliases: |
4933422O14Rik, 9630002F18, AI845682, AI848168, BB075781, D17Bwg0511e, GKAP/SAPAP, GKPA/SAPAP, Gkap, Sapap1, mKIAA4162 |
Location: |
Chr.17:69969078-70821413 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dlgap1 | NM_027712.3 | 1 | 40 | NP_081988.3 |
NM_177639.6 | 1 | 40 | NP_808307.2 | |
AK031410.1 | 5 | 730 | BAC27391.1 | |
AK033901.1 | 4 | 573 | BAC28508.1 | |
AK035763.1 | 4 | 591 | ||
AK043683.1 | 4 | 626 | ||
AK147448.1 | 5 | 857 | BAE27919.1 | |
AK220515.1 | 4 | 920 | BAD90519.1 | |
BC062120.1 | 1 | 41 | AAH62120.1 |
More Information
Set Membership: |
Intragenic Exonic |