Assay Details
Target Gene Details
Entrez Gene ID: | 224065 |
Gene Name: | urotensin 2B |
Gene Aliases: |
Gm538, U2B, Urp, Uts2d |
Location: |
Chr.16:27353322-27370239 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Uts2b | NM_198166.3 | 1 | 62 | NP_937809.1 |
AB116020.1 | BAC98928.1 | |||
AK131972.1 | 1 | 40 | BAE20912.1 | |
AK131973.1 | 1 | 43 | BAE20913.1 | |
AK132025.1 | 1 | 63 | BAE20950.1 | |
AV144685.2 | 1 | 76 | ||
AY321315.1 | 1 | 44 | AAQ83885.1 | |
BB611261.1 | 1 | 63 | ||
BC117024.1 | 1 | 34 | ||
BC117026.1 | 1 | 34 |