Assay Details
Target Gene Details
Entrez Gene ID: | 50766 |
Gene Name: | cysteine rich transmembrane BMP regulator 1 (chordin like) |
Gene Aliases: |
AU015004 |
Location: |
Chr.17:78200185-78376592 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Crim1 | NM_015800.3 | NP_056615.1 | ||
AF168680.1 | AAF34410.1 | |||
AK034889.1 | BAC28869.1 | |||
AK053197.1 | ||||
AK159603.1 | BAE35222.1 | |||
AK220139.1 | ||||
BC100348.1 | AAI00349.1 |
Target Gene Details
Entrez Gene ID: | 225020 |
Gene Name: | fasciculation and elongation protein zeta 2 (zygin II) |
Gene Aliases: |
9030616F16, D17Ertd315e |
Location: |
Chr.17:78369212-78418152 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fez2 | NM_001285949.1 | 8 | 2971 | NP_001272878.1 |
AK165595.1 | 8 | 2921 | BAE38283.1 |