Assay Details
Target Gene Details
Entrez Gene ID: | 50766 |
Gene Name: | cysteine rich transmembrane BMP regulator 1 (chordin like) |
Gene Aliases: |
AU015004 |
Location: |
Chr.17:78200185-78376592 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Crim1 | NM_015800.3 | 13 | 2887 | NP_056615.1 |
AF168680.1 | 13 | 2339 | AAF34410.1 | |
AK034889.1 | 1 | 115 | BAC28869.1 | |
AK053197.1 | 11 | 1830 | ||
AK220139.1 | 1 | 78 | ||
BC100348.1 | 13 | 2672 | AAI00349.1 |