Assay Details
Target Gene Details
Entrez Gene ID: | 13733 |
Gene Name: | adhesion G protein-coupled receptor E1 |
Gene Aliases: |
DD7A5-7, EGF-TM7, Emr1, F4/80, Gpf480, Ly71, TM7LN3 |
Location: |
Chr.17:57358651-57483529 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adgre1 | NM_010130.4 | 20 | 2596 | NP_034260.1 |
AK133202.1 | 20 | 2572 | BAE21555.1 | |
AK149898.1 | 20 | 2581 | ||
AK151303.1 | 20 | 2568 | ||
AK151682.1 | 20 | 2588 | BAE30606.1 | |
AK155862.1 | 20 | 2547 | BAE33466.1 | |
BC075688.1 | 20 | 2568 | AAH75688.1 | |
U66888.1 | 20 | 2583 | AAC53184.1 | |
X93328.1 | 20 | 2592 | CAA63720.1 |
More Information
Set Membership: |
Intragenic Exonic |