Assay Details
Target Gene Details
Entrez Gene ID: | 30940 |
Gene Name: | ubiquitin specific peptidase 25 |
Gene Aliases: |
- |
Location: |
Chr.16:77013789-77116783 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Usp25 | NM_013918.2 | NP_038946.2 | ||
AF170563.1 | AAF32264.1 | |||
AK011798.1 | BAB27849.1 | |||
AK036456.1 | BAC29438.2 | |||
BC048171.1 | AAH48171.1 | |||
BC063059.1 | AAH63059.1 |
More Information
Set Membership: |
Intragenic Non-exonic |