Assay Details
Target Gene Details
Entrez Gene ID: | 170716 |
Gene Name: | cytochrome P450, family 4, subfamily f, polypeptide 13 |
Gene Aliases: |
0610030I10Rik, Cypf13 |
Location: |
Chr.17:32924688-32947415 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyp4f13 | NM_130882.1 | NP_570952.1 | ||
AF233643.1 | AAK15009.1 | |||
AK085008.1 | BAC39336.1 | |||
BC003954.1 | AAH03954.1 |
More Information
Set Membership: |
Intragenic Non-exonic |