Assay Details
Target Gene Details
Entrez Gene ID: | 20463 |
Gene Name: | cytochrome c oxidase subunit VIIa polypeptide 2-like |
Gene Aliases: |
COX7AR, COX7RP, EB1, SIG-81, SIG81, Silg81 |
Location: |
Chr.17:83501917-83514333 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cox7a2l | NM_001159529.1 | NP_001153001.1 | ||
NM_009187.3 | NP_033213.1 | |||
AK009614.1 | ||||
AK133520.1 | BAE21701.1 | |||
AK150034.1 | BAE29256.1 | |||
AK152553.1 | BAE31307.1 | |||
AK159441.1 | BAE35087.1 | |||
AK166761.1 | BAE39000.1 | |||
AK167062.1 | BAE39224.1 | |||
AK169619.1 | ||||
AK184659.1 | ||||
AK216072.1 | ||||
AK216357.1 | ||||
BC004715.1 | AAH04715.1 | |||
BC085296.1 | AAH85296.1 | |||
BY122624.1 | ||||
X80899.1 | CAA56861.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |