Assay Details
Target Gene Details
Entrez Gene ID: | 12648 |
Gene Name: | chromodomain helicase DNA binding protein 1 |
Gene Aliases: |
4930525N21Rik, AI851787, AW555109 |
Location: |
Chr.17:15704967-15772612 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Chd1 | NM_007690.3 | 12 | 2037 | NP_031716.2 |
AK049686.1 | 12 | 2032 | BAC33872.1 | |
AK140386.1 | 12 | 2017 | BAE24366.1 | |
BC115822.1 | 11 | 1675 | ||
L10410.1 | 12 | 1714 | AAB08486.1 |