Assay Details
Target Gene Details
Entrez Gene ID: | 53817 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 39B |
Gene Aliases: |
0610030D10Rik, AI428441, Bat-1, Bat1, Bat1a, D17H6S81E, D17H6S81E-1, D6S81Eh |
Location: |
Chr.17:35241746-35253707 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx39b | NM_001252457.1 | 10 | 1344 | NP_001239386.1 |
NM_019693.3 | 10 | 1432 | NP_062667.1 | |
AA231260.1 | ||||
AF118128.1 | 9 | 1198 | AAD13115.1 | |
AK051034.1 | 10 | 1365 | BAC34505.1 | |
AK088867.1 | 10 | 1350 | BAC40624.1 | |
AK202733.1 | ||||
AK216397.1 | ||||
AY255786.1 | 9 | 1324 | AAP91685.1 | |
AY255787.1 | 9 | 1324 | AAP91686.1 | |
BC011067.1 | 10 | 1340 | AAH11067.1 | |
BC024859.1 | 9 | 2568 | AAH24859.1 |