Assay Details
Target Gene Details
Entrez Gene ID: | 224432 |
Gene Name: | SR-related CTD-associated factor 4 |
Gene Aliases: |
AA517739, Sfrs15, Sra4, Srsf15 |
Location: |
Chr.16:90228951-90284498 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Scaf4 | NM_178923.3 | NP_849254.2 | ||
AK045266.1 | ||||
AK048239.1 | 5 | 892 | BAC33280.1 | |
AK173120.1 | BAD32398.1 | |||
AK216612.1 | ||||
AK219509.1 | ||||
BC053096.1 | AAH53096.1 | |||
BC057592.1 | AAH57592.1 |
More Information
Set Membership: |
Intragenic Non-exonic |