Assay Details
Target Gene Details
Entrez Gene ID: | 51789 |
Gene Name: | tyrosine kinase, non-receptor, 2 |
Gene Aliases: |
Ack, Ack-1, Ack1, Cdgip, Pyk1 |
Location: |
Chr.16:32644099-32683493 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 16 - Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tnk2 | NM_001110147.1 | NP_001103617.1 | ||
NM_001289443.1 | NP_001276372.1 | |||
NM_016788.3 | NP_058068.2 | |||
AF037260.1 | AAC04786.1 | |||
AK087965.1 | BAC40063.1 | |||
BC031168.1 | AAH31168.1 | |||
BC052421.1 | AAH52421.1 | |||
DQ666696.1 |