Assay Details
Target Gene Details
Entrez Gene ID: | 74478 |
Gene Name: | sorting nexin 29 |
Gene Aliases: |
4933437K13Rik, ENSMUSG00000079738, Gm11170, Gm1737, Gm930, Rundc2a |
Location: |
Chr.16:11321256-11755931 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Snx29 | NM_001290148.1 | NP_001277077.1 | ||
NM_028964.4 | NP_083240.2 | |||
AF399755.1 | AAK94018.1 | |||
AK017102.1 | BAB30596.1 |
More Information
Set Membership: |
Intragenic Non-exonic |