Assay Details
Target Gene Details
Entrez Gene ID: | 20583 |
Gene Name: | snail family zinc finger 2 |
Gene Aliases: |
Slug, Slugh, Snail2 |
Location: |
Chr.16:14705859-14709385 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Snai2 | NM_011415.2 | NP_035545.1 | ||
AK133968.1 | BAE21961.1 | |||
BC062164.1 | AAH62164.1 | |||
U79550.1 | AAB38365.1 | |||
U97059.1 | AAB58704.1 |
More Information
Set Membership: |
Intragenic Non-exonic |