Assay Details
Target Gene Details
Entrez Gene ID: | 385668 |
Gene Name: | Leber congenital amaurosis 5-like |
Gene Aliases: |
4921526F01Rik, Lcca5l |
Location: |
Chr.16:96158405-96193993 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lca5l | NM_001001492.2 | NP_001001492.2 | ||
AK029554.1 | BAC26513.1 | |||
AK031481.1 | ||||
AK133316.1 |