Assay Details
Target Gene Details
Entrez Gene ID: | 26559 |
Gene Name: | hormonally upregulated Neu-associated kinase |
Gene Aliases: |
Bstk1, Mak-v |
Location: |
Chr.16:90385797-90499555 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hunk | NM_015755.2 | NP_056570.1 | ||
AF055919.2 | AAC61489.1 | |||
AF167987.1 | AAF35282.1 | |||
AK047337.1 | ||||
AK144639.1 | BAE25983.1 | |||
AK147425.1 | ||||
AK147734.1 | BAE28103.1 | |||
BC137788.1 | ||||
BC137790.1 |
More Information
Set Membership: |
Intragenic Non-exonic |