Assay Details
Target Gene Details
Entrez Gene ID: | 19876 |
Gene Name: | roundabout homolog 1 (Drosophila) |
Gene Aliases: |
AW494633, AW742721, DUTT1, Gm310 |
Location: |
Chr.16:72027553-73047491 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Robo1 | NM_019413.2 | NP_062286.2 | ||
Y17793.1 | CAA76850.1 |