Assay Details
Target Gene Details
Entrez Gene ID: | 224020 |
Gene Name: | phosphatidylinositol 4-kinase, catalytic, alpha polypeptide |
Gene Aliases: |
Pik4ca |
Location: |
Chr.16:17280351-17406314 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 50 - Exon 51 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pi4ka | NM_001001983.2 | NP_001001983.2 | ||
AK136942.1 | BAE23184.1 | |||
BC049252.1 | AAH49252.1 | |||
BC055479.1 | AAH55479.1 | |||
BC075629.1 | AAH75629.1 |
More Information
Set Membership: |
Intragenic Non-exonic |