Assay Details
Target Gene Details
Entrez Gene ID: | 15260 |
Gene Name: | histone cell cycle regulation defective homolog A (S. cerevisiae) |
Gene Aliases: |
AA138857, D16Ertd95e, Gm15797, N28177, OTTMUSG00000026264, Tuple1 |
Location: |
Chr.16:18876750-18970309 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hira | NM_010435.2 | 8 | 1507 | NP_034565.2 |
BC052856.1 | 8 | 925 | AAH52856.1 | |
X75295.1 | 9 | 1022 | CAA53043.1 | |
X92590.1 | 8 | 994 | CAA63334.1 | |
X99712.1 | 8 | 834 | CAA68049.1 |
More Information
Set Membership: |
Intragenic Exonic |