Assay Details
Target Gene Details
Entrez Gene ID: | 74143 |
Gene Name: | optic atrophy 1 |
Gene Aliases: |
1200011N24Rik, AI225888, AI847218, lilr3, mKIAA0567 |
Location: |
Chr.16:29579306-29663122 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Opa1 | NM_001199177.1 | 26 | 2817 | NP_001186106.1 |
NM_133752.3 | 25 | 2763 | NP_598513.1 | |
AB044138.1 | 25 | 2755 | BAB59000.1 | |
AK004715.1 | 26 | 2773 | ||
AK029157.1 | 25 | 2721 | BAC26331.1 | |
AK030835.1 | 26 | 2792 | ||
AK122311.1 | 25 | 2692 | BAC65593.1 | |
AK145620.1 | 25 | 2715 | BAE26544.1 | |
BC025160.1 | 7 | 779 | AAH25160.1 | |
BC138665.1 | 25 | 2644 | ||
BC145959.1 | 25 | 2644 |
More Information
Set Membership: |
Intragenic Exonic |