Assay Details
Target Gene Details
Entrez Gene ID: | 74143 |
Gene Name: | optic atrophy 1 |
Gene Aliases: |
1200011N24Rik, AI225888, AI847218, lilr3, mKIAA0567 |
Location: |
Chr.16:29579306-29663122 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Opa1 | NM_001199177.1 | NP_001186106.1 | ||
NM_133752.3 | NP_598513.1 | |||
AB044138.1 | BAB59000.1 | |||
AK004715.1 | ||||
AK029157.1 | BAC26331.1 | |||
AK030835.1 | ||||
AK122311.1 | BAC65593.1 | |||
AK145620.1 | BAE26544.1 | |||
BC025160.1 | AAH25160.1 | |||
BC138665.1 | ||||
BC145959.1 |
More Information
Set Membership: |
Intragenic Non-exonic |