Assay Details
Target Gene Details
Entrez Gene ID: | 18203 |
Gene Name: | N-terminal Asn amidase |
Gene Aliases: |
- |
Location: |
Chr.16:13818834-13835450 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ntan1 | NM_010946.3 | NP_035076.1 | ||
AK041112.1 | BAC30825.1 | |||
AK207258.1 | ||||
BC030172.1 | AAH30172.1 | |||
U57692.1 | AAB66490.1 |
Target Gene Details
Entrez Gene ID: | 94184 |
Gene Name: | pyridoxal-dependent decarboxylase domain containing 1 |
Gene Aliases: |
2210010A19Rik, AA415817, Kiaa0251-hp |
Location: |
Chr.16:13833161-13903145 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pdxdc1 | NM_001039533.2 | 23 | 2510 | NP_001034622.1 |
NM_001291017.1 | 24 | 2740 | NP_001277946.1 | |
AK004611.1 | 23 | 2387 | BAB23408.1 | |
AK054329.1 | 24 | 2723 | BAC35732.1 | |
AK163696.1 | 24 | 2611 | BAE37462.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |