Assay Details
Target Gene Details
Entrez Gene ID: | 245841 |
Gene Name: | polymerase (RNA) II (DNA directed) polypeptide H |
Gene Aliases: |
- |
Location: |
Chr.16:20717826-20722272 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Polr2h | NM_145632.2 | NP_663607.1 | ||
AK146429.1 | BAE27163.1 | |||
AK161617.1 | BAE36495.1 | |||
BC002306.1 | AAH02306.1 | |||
BC092087.1 | AAH92087.1 | |||
BC125612.1 | ||||
BC125638.1 |
More Information
Set Membership: |
Intragenic Non-exonic |