Assay Details
Target Gene Details
Entrez Gene ID: | 667772 |
Gene Name: | myosin, heavy chain 15 |
Gene Aliases: |
EG667772 |
Location: |
Chr.16:49057486-49199104 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Myh15 | NM_001166210.1 | 14 | 1463 | NP_001159682.1 |