Assay Details
Target Gene Details
Entrez Gene ID: | 77782 |
Gene Name: | polymerase (DNA directed), theta |
Gene Aliases: |
A430110D14Rik |
Location: |
Chr.16:37011742-37095419 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Polq | NM_001159369.1 | NP_001152841.1 | ||
NM_029977.2 | NP_084253.1 | |||
AY074936.1 | AAL77225.1 | |||
AY147862.1 | AAN64234.1 | |||
AY147863.1 | AAN64235.1 | |||
AY147864.1 | AAN64236.1 | |||
BC138360.1 | ||||
BC157900.1 |
More Information
Set Membership: |
Intragenic Non-exonic |