Assay Details
Target Gene Details
Entrez Gene ID: | 13840 |
Gene Name: | Eph receptor A6 |
Gene Aliases: |
Ehk2, Hek12, m-ehk2 |
Location: |
Chr.16:59653061-60606484 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Epha6 | NM_007938.2 | NP_031964.2 | ||
AK032436.1 | BAC27868.1 | |||
BC141090.1 | ||||
U58332.1 | AAB53836.1 |
More Information
Set Membership: |
Intragenic Non-exonic |