Assay Details
Target Gene Details
Entrez Gene ID: | 71756 |
Gene Name: | carboxypeptidase N, polypeptide 2 |
Gene Aliases: |
1300018K11Rik |
Location: |
Chr.16:30256378-30267614 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cpn2 | NM_027904.3 | NP_082180.2 | ||
AK005049.1 | BAB23775.1 | |||
BC138287.1 | 1 | 46 | ||
BC138288.1 | 1 | 46 |
More Information
Set Membership: |
Intragenic Non-exonic |