Assay Details
Target Gene Details
Entrez Gene ID: | 224129 |
Gene Name: | adenylate cyclase 5 |
Gene Aliases: |
AW121902, Ac5 |
Location: |
Chr.16:35155636-35304549 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adcy5 | NM_001012765.4 | NP_001012783.3 | ||
AK160942.1 | BAE36104.1 | |||
BC090846.1 | AAH90846.1 |